Mouse Interferon gamma ELISA Kit
Supplier: Abcam
AB46081-96 AB46081-2X96
ABCAAB46081-96EA 969 EUR
ABCAAB46081-96 ABCAAB46081-2X96
Mouse Interferon gamma ELISA Kit
Assays ELISAs
Mouse Interferon gamma ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Interferon gamma in Mouse in Cell culture supernatant, Serum samples.