TriLink® CleanAmp® dNTP Mix
Supplier: TriLink BioTechnologies
N-9506-10 N-9506-2
SOLUN-9506-10EA 466 EUR
SOLUN-9506-10 SOLUN-9506-2
TriLink® CleanAmp® dNTP Mix
Nucleic Acid Reagents Nucleotides
CleanAmp® dNTPs are chemically-modified nucleotides designed to improve specificity, sensitivity, and robustness of your PCR.
  • Improves PCR specificity, sensitivity, and robustness
  • Provides an alternative approach to hot-start PCR without a need for a specially designed DNA polymerase
  • Grade: RUO
The CleanAmp® dNTP Mix: - Contains 10 mM each of CleanAmp® dATP, dCTP, dGTP, and dTTP - Enables hot-start PCR for specificity and sensitivity, with a thermolabile protecting group at the 3’ hydroxyl of the nucleotides (CleanAmp® dNTP benefit) - Has been QC analyzed by AX-HPLC, RP-HPLC, 31P NMR, mass spec, and a standardised PCR assay
Caution: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eed637cea91c5c517ced4ff91e67f48cee5d0c4a&vid=eed637cea91c5c517ced4ff91e67f48cee5d0c4a&prid=web&clid=SAPDAM
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable
Application
Learn more