Fixed Angle Rotors for MIKRO 2.0 Centrifuges
Supplier: Hettich
1540 2413 2414 2415 2419
521-4356EA 895 EUR
521-4356 521-4358 521-4354 521-4355 521-4357
Fixed Angle Rotors for MIKRO 2.0 Centrifuges
Centrifuges Rotors
Angle rotors, 45°, with lid bioseal. 
  • Quick-release lid closure
The quick-release lid closure improves handling by allowing each rotor to be quickly and biologically sealed, opened, and safely transported. This facilitates handling and speeds up the centrifugation process.