EMBER™ Ultra DNA Gel Stain Kit
Supplier: Biotium
41043 41043-T
41043.EA 589 EUR
41043. 41043-T
EMBER™ Ultra DNA Gel Stain Kit
Stains Electrophoresis Stains
The most sensitive DNA gel stain available in convenient pre-coated agarose format with no need for post-electrophoresis staining.
  • Easy-to-use pre-coated agarose format for DNA gel electrophoresis
  • Superior sensitivity, detect as little as ≤1 ng DNA
  • Optimal for blue LED gel imagers
  • Also compatible with UV imagers
The EMBER™ Ultra DNA gel system provides the highest sensitivity and resolution for DNA. Simply cast the agarose gel using the EMBER™ Ultra precoated agarose and prepare the samples in the EMBER™ Ultra DNA loading dye. Bands can be imaged immediately after electrophoresis.
Note: The number of gels per kit is based on a 90×110 mm, 1% agarose mini gel (0,5 g agarose in 50 ml buffer per gel). The actual number may vary depending on gel size.
EMBER™ Ultra Precoated Agarose is ultra-pure molecular biology grade agarose with low electroendosmosis that is precoated with Biotium’s fluorescent EMBER™ Ultra Dye.This kit helps in detection of as little as ≤1 ng DNA.
The gel system does not require a post-electrophoresis staining step, for convenient and fast results. EMBER™ Ultra Precoated Agarose gives excellent results at percentages between 1% and 5% agarose, for analysing different sizes of DNA. EMBER™ Ultra also has excellent sensitivity for single-stranded DNA and small DNA fragments.
The loading dye has green fluorescence with excitation/emission at 500/530 nm when bound to nucleic acids. The loading dye contains blue and orange tracking dyes that migrate at ~1500 bp (blue) and ~50 bp (orange) in a 1% agarose gel.
Delivery information:
Kit components: EMBER™ Ultra precoated agarose, EMBER™ Ultra DNA loading dye.