EasySep™ Easy 250 Magnet
Supplier: STEMCELL Technologies
100-0821
STMC100-0821EA 0 EUR
STMC100-0821
EasySep™ Easy 250 Magnet
Magnets
Scale up manual cell isolations from large-volume samples, such as leukopaks and whole blood. Rather than splitting cell suspensions and performing multiple rounds of isolations, the Easy 250 magnet can process up to 225 ml and 1.25×10¹⁰ cells in a single isolation.
  • Magnet for column-free immunomagnetic separation
  • Increase cell isolation throughput by isolating cells from large-volume samples in a single separation
  • Isolate immune cells from samples up to 225 ml in as little as 20 minutes
  • Achieve up to 99% cell purities of viable and functional cells
  • Obtain