EasySep™ Human ILC2 Isolation Kit
Supplier: STEMCELL Technologies
17782
STMC17782EA 1950 EUR
STMC17782
EasySep™ Human ILC2 Isolation Kit
Cell Separation Media
Immunomagnetic cell isolation kit using particle release technology.
  • Easy-to-use and column-free
  • Up to 95% purity
  • Facilitates rapid isolation of ILC2s without flow sorting
  • No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Human ILC2 Isolation Kit is designed to isolate Group 2 Innate Lymphoid Cells (ILC2s) from leukapheresis products by immunomagnetic positive selection without the use of a flow sorter.
First, CRTH2+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ ReleasableMore Product Information