EasySep™ Human Extracellular Vesicle (CD63) Positive Selection Kit
Supplier: STEMCELL Technologies
17895
STMC17895EA 575 EUR
STMC17895
EasySep™ Human Extracellular Vesicle (CD63) Positive Selection Kit
Cell Separation Media
Fast and easy isolation of human extracellular vesicles using immunomagnetic positive selection.
  • Fast, easy to use, and column-free
  • Avoid the need for ultracentrifugation and associated time consuming EV isolation methods
  • Obtain highly purified EVs without contaminating biofluid components
The EasySep™ Human Extracellular Vesicle (CD63) Positive Selection Kit isolates highly purified human extracellular vesicles from plasma/serum and cell culture conditioned medium by immunomagnetic positive selection. Extracellular vesicles are labelled withMore Product Information