EasySep™ Magnet
Supplier: STEMCELL Technologies
18000
STMC18000EA 2180 EUR
STMC18000
EasySep™ Magnet
Magnets
The EasySep™ magnet is designed for cell separation procedures using EasySep™ reagents.
  • Immunomagnetic column-free magnet
  • Designed to hold a standard 12×75 mm (5 ml) polystyrene tube
The EasySep™ magnet generates a high-gradient magnetic field in the interior cavity that is strong enough to separate cells labeled with EasySep™ magnetic particles, without the use of columns.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available forMore Product Information