Anti-CD25 Rat Monoclonal Antibody [clone: PC61.5] (PerCP-Cy5.5)
Supplier: STEMCELL Technologies
60009PS 60009PS.1
STMC60009PSEA 655 EUR
STMC60009PS STMC60009PS.1
Anti-CD25 Rat Monoclonal Antibody [clone: PC61.5] (PerCP-Cy5.5)
Antibodies
Mouse monoclonal IgG1 antibody against human CD34, FITC-conjugated.
  • Hematopoietic stem
  • Progenitor cells
The 581 antibody reacts with human CD34, an ~105 to 120 kDa type 1 transmembrane glycoprotein expressed on the surface of most human hematopoietic stem and progenitor cells (HSPCs) as well as on mesenchymal stem cells, embryonic fibroblasts, endothelial cells, neurons, and some tumor cell lines. CD34 is expressed only transiently during hematopoiesis, so the frequency of CD34+ cells is low in bone marrow or cord blood (~1 to 5%) and very low (~0.1More Product Information