Anti-CD32 Mouse Monoclonal Antibody [clone: IV.3]
Supplier: STEMCELL Technologies
60012
STMC60012EA 426 EUR
STMC60012
Anti-CD32 Mouse Monoclonal Antibody [clone: IV.3]
Antibodies
Mouse monoclonal IgG2b antibody against human CD32, unconjugated.
  • B cells
  • Granulocytes
  • Subsets
  • Monocytes
The IV.3 antibody reacts with human CD32 (FcγRII), an ~40 kDa type 1 transmembrane glycoprotein that mediates several functions including phagocytosis, cytotoxicity, immunomodulation and platelet aggregation. CD32 is encoded by three genes (A, B, C) and at least 6 isoforms are generated via alternative mRNA splicing, i.e., IIa1, IIa2, IIb1, IIb2, IIb3 and IIc. All isoforms are expressed by monocytes/macrophages, placental trophoblasts andMore Product Information