Anti-CD3e Hamster Monoclonal Antibody [clone: 145-2C11] (Alexa Fluor® 488)
Supplier: STEMCELL Technologies
60015AD
STMC60015ADEA 457 EUR
STMC60015AD
Anti-CD3e Hamster Monoclonal Antibody [clone: 145-2C11] (Alexa Fluor® 488)
Antibodies
Hamster (Armenian) monoclonal IgG1 antibody against mouse CD3e, Alexa Fluor® 488-conjugated.
  • T cells
The 145-2C11 antibody reacts with the ~20 kDa CD3ε subunit of the mouse T cell receptor (TCR)/CD3 complex, which is expressed on the surface of circulating mature T cells and NKT cells, and variably on thymocytes. A majority of T cell neoplasms also express CD3. The CD3 complex, which is assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, associates non-covalently with the TCR and is involved in transducing antigen recognitionMore Product Information