Anti-Igm Isotype Control Rat Monoclonal Antibody [clone: RTK2118] (Alexa Fluor® 488)
Supplier: STEMCELL Technologies
60074AD
STMC60074ADEA 212 EUR
STMC60074AD
Anti-Igm Isotype Control Rat Monoclonal Antibody [clone: RTK2118] (Alexa Fluor® 488)
Antibodies
Rat monoclonal IgM, kappa isotype control antibody, Alexa Fluor® 488-conjugated.
The RTK2118 antibody (IgM, kappa) is suitable for use as an isotype-matched control antibody in several applications to estimate the degree of non-specific binding by an antigen-specific antibody. Ideally, the isotype control should have the same subclass of heavy chain (IgA, IgD, IgE, IgG, or IgM) and light chain (kappa or lambda) as the specific antibody being used. If a conjugated antibody is used, an isotype control conjugated to the same molecule (e.g. fluorochrome) should be chosen. The use of anMore Product Information