Anti-CD24 Rat Monoclonal Antibody [clone: M1/69] (PerCP-Cy5.5)
Supplier: STEMCELL Technologies
60099PS 60099PS.1
STMC60099PSEA 681 EUR
STMC60099PS STMC60099PS.1
Anti-CD24 Rat Monoclonal Antibody [clone: M1/69] (PerCP-Cy5.5)
Antibodies
Rat monoclonal IgG2b antibody against mouse CD24, PerCP-Cy5.5-conjugated.
  • B cells
  • Mammary cells
The M1/69 antibody reacts with murine CD24, an ~35 - 45 kDa glycoprotein expressed on lymphocytes, granulocytes, thymocytes, erythrocytes, epithelial cells, neurons, and dendritic cells. CD24 is differentially expressed during T and B cell development, and is not found on the majority of mature peripheral T cells or plasma cells. It is expressed on pro-B, pre-B and mature B cells, but expression is strongly down-regulated after B cell activation. CD24 isMore Product Information