Mouse Recombinant TPO
Supplier: STEMCELL Technologies
78072 78072.1 78072.2
STMC78072EA 694 EUR
STMC78072 STMC78072.1 STMC78072.2
Mouse Recombinant TPO
Proteins and Peptides
Thrombopoietin (TPO) is a key regulator of megakaryocytopoiesis and thrombopoiesis in vitro and in vivo. TPO stimulates the proliferation and maturation of megakaryocytes and has an important role in regulating the level of circulating platelets in vivo. TPO also promotes the survival, self-renewal, and expansion of hematopoietic stem cells and primitive multilineage progenitor cells. It is commonly used with other cytokines such as stem cell factor (SCF) and Flt3/Flk-2 Ligand to promote expansion of primitive hematopoietic cells in culture (Hitchcock andamp; Kaushansky). The TPO receptor, c-Mpl, is expressed at all stages of megakaryopoiesis, from hematopoietic stem and progenitor cells to mature platelets.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').