Human Recombinant VEGF-D
Supplier: STEMCELL Technologies
78203 78203.1
STMC78203EA 276 EUR
STMC78203 STMC78203.1
Human Recombinant VEGF-D
Proteins and Peptides
Vascular endothelial growth factor D (VEGF-D) is a member of the VEGF/platelet-derived growth factor (PDGF) family of proteins. VEGF-D is a potent angiogenic factor and promotes lymphangiogenesis, endothelial cell growth and survival, and can affect blood vessel permeability. VEGF-D is expressed in the lung, heart, small intestine, fetal lung, and at lower levels in the pancreas, colon, and skeletal muscle (Otrock et al.; Roy et al.; Stacker et al.; Yamada et al.). VEGF-D is a ligand for VEGF receptors 2 (Flk1) and 3 (Flt4) (Baldwin et al.). VEFGR-3 is highly expressed in lymphatic endothelial cells and is essential for their growth and differentiation (Otrock et al.; Roy et al.). Binding of VEGF-D to neuropilins contributes to VEGFR-3 signaling during lymphangiogenesis, whereas binding to integrin α9β1 promotes endothelial cell adhesion and migration (Roy et al.; Otrock et al.). During embryogenesis, VEGF-D also plays a role in the formation of the venous and lymphatic systems.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').