Anti-GFAP Mouse Monoclonal Antibody [clone: 2E1.E9]

Supplier: STEMCELL Technologies
60048 60048.1
STMC60048EA 590 EUR
STMC60048 STMC60048.1
Anti-GFAP Mouse Monoclonal Antibody [clone: 2E1.E9]
Antibodies
Mouse monoclonal IgG2b antibody against human, mouse, rat GFAP (glial fibrillary acidic protein), unconjugated.

  • Astrocytes
  • Neural cells Psc Derived

The 2E1.E9 antibody reacts with glial fibrillary acidic protein (GFAP), an ~49 kDa type III intermediate filament (IF) protein that, within the central nervous system, is expressed primarily by astrocytes, though found at high levels in some glial-derived tumors. GFAP is thought to contribute to the structural architecture and strength of the cytoskeleton. The 2E1.E9 antibody does not cross-react with other IF proteins and can be used to distinguish astrocytes from other glial cells. GFAP has also been identified in Leydig cells, keratinocytes, chondrocytes and osteocytes. The GFAP polypeptide comprises an N-terminal head, a central rod, and a C-terminal tail domain, and assembles as dimers by a process dependent on phosphorylation and dephosphorylation of the N-terminal domain. Several splice variants have been identified, encoding three distinct isoforms. Many mutations in the GFAP gene (>50) have been associated with Alexander disease, a progressive leukoencephalopathy characterized by cytoplasmic inclusions and dysfunctional myelination. This antibody clone has been verified for labeling neural stem and progenitor cells grown in NeuroCult™ NS-A Proliferation Kit (Human; Catalog #05751) and NeuroCult™ Proliferation Kit (Mouse; Catalog #05702).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type:
Antigen: GFAP
Clonality: Monoclonal
Clone: 2E1.E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR